
2′,4′,6′-Trihydroxyacetophenone monohydrate

Synonym(s):
2-Acetylphloroglucinol, THAP
Linear Formula:
(HO)3C6H2COCH3·H2O
CAS Number:
Molecular Weight:
186.16
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Analyte Suitability
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing