
2,3-Dichlorophenol

Synonym(s):
2,3-Dichlorophenol
Linear Formula:
2,3-(Cl)2C6H3OH
CAS Number:
Molecular Weight:
163.00
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing