Skip to Content
BA
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
19-Nortestosterone
Synonym(s):
17β-Hydroxy-19-norandrost-4-en-3-one, 17β-Hydroxy-4-estren-3-one, 19-Norandrostenolone, 4-Estren-17β-ol-3-one, Nadrolone, Nandrolone
Empirical Formula (Hill Notation):
C
18
H
26
O
2
CAS Number:
434-22-0
Molecular Weight:
274.40
EC Number:
207-101-0
Products
Product Category
bioactive small molecules (1)
building blocks (1)
hormones (1)
Brand
Sigma-Aldrich (2)
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Markush Class
Markush Group
Special Grade
Puriss (1)
Sterility
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
N7252
19-Nortestosterone,
Expand
View Pricing
74640
19-Nortestosterone,
≥99.0% (HPLC)
Expand
View Pricing