Skip to Content
BA
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,3-Diolein
Synonym(s):
1,3-Di(cis-9-octadecenoyl)glycerol
Empirical Formula (Hill Notation):
C
39
H
72
O
5
CAS Number:
2465-32-9
Molecular Weight:
620.99
EC Number:
219-569-3
Products
Product Category
glycerides (1)
lipids (1)
metabolites (1)
Brand
Sigma-Aldrich (1)
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
D3627
1,3-Diolein,
≥99% (GC)
Expand
View Pricing