Skip to Content
Merck
All Photos(1)

Key Documents

SML0504

Sigma-Aldrich

I-BOP

≥95% (HPLC)

Synonym(s):

(5Z)-7-[(1S,2R,3R,4R)-3-[(1E,3R)-3-Hydroxy-4-(4-iodophenoxy)-1-buten-1-yl]-7-oxabicyclo[2.2.1]hept-2-yl]-5-heptenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H29IO5
CAS Number:
Molecular Weight:
512.38
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solution

concentration

10 mg/mL in ethanol

color

colorless to faint yellow

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

I-BOP is a potent thromboxane (TP) receptor agonist. I-BOP induces human platelet aggregation with an EC50 value of 0.34 nM.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service