Skip to Content
Merck
All Photos(1)

Key Documents

SAB2900089

Sigma-Aldrich

Anti-SLC7A5 (AB2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

4F2 light chain, 4F2LC, CD98 light chain, CD98LC, D0H16S474E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

monkey, human, dog, rabbit

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SLC7A5(8140)

Related Categories

Immunogen

Synthetic 14 amino acid peptide from N-terminus of human SLC7A5. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset, Dog, Elephant, Rabbit, Platypus (100%); Opossum (93%); Mouse, Rat, Bat, Hamster, Panda, Chicken (86%).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masato Asanuma et al.
PloS one, 9(9), e106362-e106362 (2014-09-05)
L-DOPA is therapeutically efficacious in patients with Parkinson's disease (PD), although dopamine (DA) neurons are severely degenerated. Since cortical astrocytes express neutral amino acid transporter (LAT) and DA transporter (DAT), the uptake and metabolism of L-DOPA and DA in striatal
Simona Infantino et al.
Science signaling, 7(338), ra77-ra77 (2014-08-15)
Maintenance of an appropriate number of plasma cells, long-lived antibody-producing cells that are derived from B cells, is essential for maintaining immunological memory while limiting disease. Plasma cell survival relies on extrinsic factors, the limited availability of which determines the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service