Skip to Content
Merck
All Photos(1)

Key Documents

NP7

Sigma-Aldrich

TERGITOL

Type NP-7

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900

description

non-ionic

type

Type NP-7

mol wt

528 g/mol

CMC

39 ppm

transition temp

cloud point 20 °C

HLB

12

SMILES string

CCCCCCCCCC1=CC=C(C=C1)OCCO

InChI

1S/C17H28O2/c1-2-3-4-5-6-7-8-9-16-10-12-17(13-11-16)19-15-14-18/h10-13,18H,2-9,14-15H2,1H3

InChI key

KUXGUCNZFCVULO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tergitol is a non-ionic surfactant.

Application

Tergitol has been used in a study to develop a method for the determination of phenolic compounds in environmental water samples. It has also been used in a study to investigate the structural characterization and antimicrobial properties of silver nanoparticles.

Other Notes

Polyglycol ether (nonionic) surfactants.

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.5 °F - Pensky-Martens closed cup

Flash Point(C)

110.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Huihui Ma et al.
Journal of separation science, 35(18), 2484-2490 (2012-08-30)
A method based on ultrasound-assisted cloud point extraction (CPE) was developed for the determination of phenolic compounds in water samples by high-performance liquid chromatography coupled with diode array detection (DAD) (HPLC-DAD). A nonionic surfactant, Tergitol 15-S-7, was chosen as the
Irshad A Wani et al.
Colloids and surfaces. B, Biointerfaces, 101, 243-250 (2012-09-27)
Silver nanoparticles have been synthesized in the inverse microemulsions formed using three different surfactants viz., cetyl-trimethyl ammonium bromide (CTAB), Tergitol and Triton X-100. We have done a systematic study of the effect of the surfactants on the particle size and
Dayna M Brichta-Harhay et al.
Applied and environmental microbiology, 74(20), 6289-6297 (2008-08-30)
The hide and carcass hygiene of cull cattle at slaughter in four geographically distant regions of the United States was examined from July 2005 to April 2006 by measuring the aerobic plate counts (APC) and the prevalences and loads of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service