Skip to Content
Merck
All Photos(1)

Key Documents

92342

Supelco

Galangin

analytical standard

Synonym(s):

3,5,7-Trihydroxyflavone, Norizalpinin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
272179
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

form

powder or crystals

shelf life

limited shelf life, expiry date on the label

color

faint yellow to dark yellow

mp

214-215 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

Oc1cc(O)c2C(=O)C(O)=C(Oc2c1)c3ccccc3

InChI

1S/C15H10O5/c16-9-6-10(17)12-11(7-9)20-15(14(19)13(12)18)8-4-2-1-3-5-8/h1-7,16-17,19H

InChI key

VCCRNZQBSJXYJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Galangin is a polyphenolic compound belonging to the class of flavonoids and is primarily derived from different medicinal herbs. It exhibits antiproliferative, antiangiogenic, antioxidant, anti-cancer, antiinflammatory effects and muscle contraction inhibitor properties.

Application

Galangin may be used as an analytical reference standard for the quantification of the analyte in propolis and wine samples using high-performance liquid chromatography technique.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyphenols: Prevention and Treatment of Human Disease (2018)
Determination of red wine flavonoids by HPLC and effect of aging
Fang F, et al.
Food Chemistry, 101(1), 428-433 (2007)
Analysis of flavonoids from propolis by on-line HPLC?electrospray mass spectrometry
Volpi N and Bergonzini G
Journal of Pharmaceutical and Biomedical Analysis, 42(3), 354-361 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service