Skip to Content
Merck
All Photos(1)

Key Documents

60737

Supelco

Silica gel Inorganic Sorbent

high-purity grade, 230-400 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
SB.52

product name

Silica gel, high-purity grade, pore size 60 Å, 230-400 mesh particle size, 40-63 μm particle size, for flash chromatography

grade

high-purity grade

Quality Level

form

powder

manufacturer/tradename

Sigma-Aldrich

technique(s)

LPLC: suitable

surface area

530 m2/g

impurities

3-7% water

matrix

Silica

matrix active group

silica

particle size

230-400 mesh
40-63 μm

pore size

0.8 cm3/g pore volume
60 Å

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium (III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is classified as below:
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent removed by supercritical extraction

Application


  • Modulating the properties and structure of lignins produced by alkaline delignification of sugarcane bagasse pretreated with two different mineral acids at pilot-scale: Demonstrates the use of high-purity sulfuric acid in the pretreatment of sugarcane bagasse, crucial for enhancing the quality of lignin extracted for further biochemical applications, pivotal for research in material science and environmental sustainability (Nunes da Silva et al., 2024).



Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhys B Murphy et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 19(10), 1410-1422 (2020-09-24)
The remarkable properties of deuterium have led to many exciting and favourable results in enhancing material properties, for applications in the physical, medical, and biological sciences. Deuterated isotopologues of avobenzone, a sunscreen active ingredient, were synthesised to examine for any
Junfeng Wang et al.
Journal of medicinal chemistry, 63(6), 3381-3389 (2020-02-23)
We have synthesized and characterized [18F]-N-(4-chloro-3-((fluoromethyl-d2)thio)phenyl)-picolinamide ([18F]15) as a potential ligand for the positron emission tomography (PET) imaging of mGluR4 in the brain. Radioligand [18F]15 displays central nervous system drug-like properties, including mGluR4 affinity, potent mGluR4 PAM activity, and selectivity
Gengyang Yuan et al.
Journal of medicinal chemistry, 63(20), 12060-12072 (2020-09-29)
Three benzimidazole derivatives (13-15) have been synthetized as potential positron emission tomography (PET) imaging ligands for mGluR2 in the brain. Of these compounds, 13 exhibits potent binding affinity (IC50 = 7.6 ± 0.9 nM), positive allosteric modulator (PAM) activity (EC50
Mattia Bartoli et al.
Waste management & research : the journal of the International Solid Wastes and Public Cleansing Association, ISWA, 38(2), 213-225 (2019-08-15)
Olive pruning residue is largely formed during cultivation, and is usually disposed through open-air combustion directly in the field, but this habit is a possible source of pollution. The pyrolytic conversion of olive pruning residue has been run in a
Christopher A Dyke et al.
Journal of the American Chemical Society, 127(12), 4497-4509 (2005-03-24)
In this report, procedures are discussed for the enrichment of single-walled carbon nanotube (SWNT) types by simple filtration of the functionalized SWNTs through silica gel. This separation uses nanotube sidewall functionalization employing two different strategies. In the first approach, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service