Skip to Content
Merck
All Photos(3)

Key Documents

60588

Supelco

Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate

Selectophore

Synonym(s):

Tetrakis[3,5-bis(trifluoromethyl)phenyl]boron potassium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H12BF24K
CAS Number:
Molecular Weight:
902.31
Beilstein:
8893801
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥95.0% (based on K, ICP)

mp

126-129 °C

SMILES string

[K+].FC(F)(F)c1cc(cc(c1)C(F)(F)F)[B-](c2cc(cc(c2)C(F)(F)F)C(F)(F)F)(c3cc(cc(c3)C(F)(F)F)C(F)(F)F)c4cc(cc(c4)C(F)(F)F)C(F)(F)F

InChI

1S/C32H12BF24.K/c34-25(35,36)13-1-14(26(37,38)39)6-21(5-13)33(22-7-15(27(40,41)42)2-16(8-22)28(43,44)45,23-9-17(29(46,47)48)3-18(10-23)30(49,50)51)24-11-19(31(52,53)54)4-20(12-24)32(55,56)57;/h1-12H;/q-1;+1

InChI key

DGEVYOMQOPNBCY-UHFFFAOYSA-N

General description

Potassium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate was used as one of the membrane component during preparing ion-selective electrodes (ISEs), in an experimental study conducted to classify beverages using a reduced sensor array.
Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kevin J Cash et al.
Sensors (Basel, Switzerland), 12(9), 11922-11932 (2012-11-01)
In this communication we discuss the development of ionophore based nanosensors for the detection and monitoring of histamine levels in vivo. This approach is based on the use of an amine-reactive, broad spectrum ionophore which is capable of recognizing and
Classification of beverages using a reduced sensor array.
Ciosek, Patrycja, Zbigniew Brzozka, and Wojciech Wroblewski.
Sensors and Actuators B, Chemical, 103 (1), 76-83 (2004)
Mohsen M Zareh
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 24(7), 889-894 (2008-07-11)
TFPB was introduced as a charged ionophore for atropine selective electrodes. Typical Nernstian responses were found (57.78, 58.95 and 58.41 mV/decade) for PVC-membrane electrodes incorporating NPOE, DOS, and DDP as plasticizers. They exhibited practical linear ranges of 9.1 x 10(-3)
Tanji Yin et al.
Analytica chimica acta, 1101, 50-57 (2020-02-08)
The dissolution kinetics of silver nanoparticles (Ag NPs) to Ag+ ions is a critical factor determining the toxicity of silver nanoparticles. In this work, a solid-contact Ag+-selective electrode (Ag+-ISE) is fabricated and used to monitor the dissolution of Ag NPs.
Xiaohui Chen et al.
Journal of experimental botany, 69(21), 5279-5291 (2018-08-14)
To develop elite crops with low cadmium (Cd), a fundamental understanding of the mechanism of Cd uptake by crop roots is necessary. Here, a new mechanism for Cd2+ entry into rice root cells was investigated. The results showed that Cd2+

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service