Skip to Content
Merck
All Photos(1)

Key Documents

471348

Sigma-Aldrich

Methanesulfonic acid solution

70 wt. % in H2O

Synonym(s):

Mesylic acid, MsOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO3H
CAS Number:
Molecular Weight:
96.11
Beilstein:
1446024
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.3 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

concentration

70 wt. % in H2O

impurities

≤1,000 ppm Sulfate (SO4)
≤10 ppm Iron
≤50 ppm Chloride (Cl)

refractive index

n20/D 1.416

density

1.319 g/mL at 25 °C

SMILES string

CS(O)(=O)=O

InChI

1S/CH4O3S/c1-5(2,3)4/h1H3,(H,2,3,4)

InChI key

AFVFQIVMOAPDHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The product is a 70% aqueous solution of methanesulfonic acid (MSA). MSA is a strong organic acid. The chemical oxidation of dimetyl sulfide in the atmosphere leads to the formation of MSA in large quantities. MSA undergoes biodegradation by forming CO2 and sulphate. It is considered a green acid as it is less toxic and corrosive in comparison to mineral acids.1 The aqueous MSA solution has been considered a model electrolyte for electrochemical processes.

Application

Methanesulfonic acid solution may be used to study the corrosion behavior of stainless steel.

Legal Information

Product of Arkema Inc.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Corrosion behaviour of stainless steels in aqueous solutions of methanesulfonic acid.
Finsgar M and Milosev I.
Corrosion Science, 52(7), 2430-2438 (2010)
Methane Sulphonic Acid is Green Catalyst in Organic Synthesis.
Kulkarni P.
Orient. J. Chem., 31(1), 447-451 (2015)
Microbial metabolism of methanesulfonic acid.
Kelly DP and Murrell JC.
Archives of Microbiology, 172(6), 341-348 (1999)
Environmental benefits of methanesulfonic acid. Comparative properties and advantages.
Gernon MD.
Green Chemistry, 1(3), 127-140 (1999)
Renata Solarska et al.
Nanoscale, 4(5), 1553-1556 (2012-02-01)
Nanostructuring of semiconductor films offers the potential means for producing photoelectrodes with improved minority charge carrier collection. Crucial to the effective operation of the photoelectrode is also the choice of a suitable electrolyte. The behaviour of the nanostructured WO(3) photoanodes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service