Skip to Content
Merck
All Photos(1)

Key Documents

45855

Supelco

(−)-Bornyl acetate

analytical standard

Synonym(s):

endo-(1S)-1,7,7-Trimethylbicyclo[2.2.1]hept-2-yl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29
Beilstein:
2502036
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

0.6 mmHg ( 20 °C)

Assay

≥99.0% (sum of enantiomers, GC)

optical activity

[α]20/D −46±2°, c = 1% in ethanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n/D 1.463 (lit.)
n20/D 1.463 (lit.)

bp

223-224 °C (lit.)

density

0.986 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C

InChI

1S/C12H20O2/c1-8(13)14-10-7-9-5-6-12(10,4)11(9,2)3/h9-10H,5-7H2,1-4H3/t9-,10+,12+/m0/s1

InChI key

KGEKLUUHTZCSIP-HOSYDEDBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: achillea echinacea salvia thymus valeriana

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

185.0 - 194.0 °F

Flash Point(C)

85 - 90 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bénédicta G H Kpadonou Kpoviessi et al.
Journal of ethnopharmacology, 155(3), 1417-1423 (2014-07-25)
Different parts of Ocimum gratissimum Linn are largely used in folk medicine for the treatment of many diseases, some of which related to parasitical infections as fevers and headaches. In order to validate their use and to clarify the plant
Barbara Sgorbini et al.
Journal of chromatography. A, 1376, 9-17 (2014-12-30)
Herbs and spices are used worldwide as food flavoring, thus determination of their identity, origin, and quality is mandatory for safe human consumption. An analysis strategy based on separative (HS-SPME-GC-MS) and non-separative (HS-SPME-MS) approaches is proposed for the volatile fraction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service