Skip to Content
Merck
All Photos(1)

Key Documents

36685

Supelco

2-Aminopyridine

PESTANAL®, analytical standard

Synonym(s):

2-AP, 2-Pyridinamine, 2-Pyridylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
Beilstein:
105785
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

98-100% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

204-210 °C (lit.)

mp

54-58 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1ccccn1

InChI

1S/C5H6N2/c6-5-3-1-2-4-7-5/h1-4H,(H2,6,7)

InChI key

ICSNLGPSRYBMBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Aminopyridine can be called as structural motifs, which can be encountered in a variety of useful compounds, that includes fluorescent organic materials and therapeutics. It can undergo condensation with an aromatic aldehyde to give the corresponding schiff′s base.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd?PEPPSI?IPentCl: A Useful Catalyst for the Coupling of 2?Aminopyridine Derivatives
Khadra A, et al.
Chemistry?A European Journal , 23, 3206-3212 (2017)
A novel approach towards design, synthesis and evaluation of some Schiff base analogues of 2-aminopyridine and 2-aminobezothiazole against hepatocellular carcinoma
Chacko S and Samanta S
Biomedicine and Pharmacotherapy, 89, 162-176 (2017)
Thomas Zengeya et al.
Angewandte Chemie (International ed. in English), 51(50), 12593-12596 (2012-11-06)
Peptide nucleic acids containing thymidine and 2-aminopyridine (M) nucleobases form stable and sequence-selective triple helices with double-stranded RNA at physiologically relevant conditions. The M-modified PNA showed unique RNA selectivity by having two orders of magnitude higher affinity for the double-stranded
Daniela P Gouvêa et al.
Ultrasonics sonochemistry, 19(6), 1127-1131 (2012-04-10)
The efficient multicomponent synthesis of thiazolidinones from the reaction of arenealdehydes, mercaptoacetic acid and 2-picolilamine or 2-aminopyridine under ultrasound irradiation are reported. The reaction with 2-aminopyridine needs a Lewis acid catalysis to afford the corresponding 2-aryl-3-(pyridin-2-yl)-1,3-thiazolidin-4-ones. All novel compounds were
Honggen Wang et al.
Journal of the American Chemical Society, 132(38), 13217-13219 (2010-09-09)
A novel and efficient synthesis of pyrido[1,2-a]benzimidazoles through direct intramolecular aromatic C-H amination of N-aryl-2-aminopyridines has been developed. The reaction, cocatalyzed by Cu(OAc)(2) and Fe(NO(3))(3)·9H(2)O, is carried out in DMF under a dioxygen atmosphere. Diversified pyrido[1,2-a]benzimidazoles containing various substitution patterns

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service