Skip to Content
Merck
All Photos(1)

Key Documents

34069

Supelco

Maduramicin ammonium

VETRANAL®, analytical standard

Synonym(s):

Maduramycin ammonium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H83NO17
CAS Number:
Molecular Weight:
934.16
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

[NH4+].[H][C@@]1(C[C@H](O[C@@H]2C[C@H](OC)[C@@H](OC)[C@H](C)O2)[C@@]([H])(O1)[C@]3(C)CC[C@@]([H])(O3)[C@]4(C)CC[C@]5(C[C@H](O)[C@@H](C)[C@]([H])(O5)[C@@H](C)[C@]6([H])O[C@](O)(CC([O-])=O)[C@@H](C)[C@H](OC)[C@H]6OC)O4)[C@@]7([H])O[C@](C)(O)[C@H](C)C[C@@H]7C

InChI

1S/C47H80O17.H3N/c1-23-18-24(2)45(9,51)61-36(23)31-19-32(58-35-20-30(53-10)40(55-12)28(6)57-35)42(59-31)44(8)15-14-33(60-44)43(7)16-17-46(64-43)21-29(48)25(3)37(62-46)26(4)38-41(56-13)39(54-11)27(5)47(52,63-38)22-34(49)50;/h23-33,35-42,48,51-52H,14-22H2,1-13H3,(H,49,50);1H3/t23-,24+,25+,26+,27-,28-,29-,30-,31+,32-,33+,35+,36-,37-,38-,39-,40-,41-,42+,43-,44-,45-,46+,47+;/m0./s1

InChI key

WQGJEAMPBSZCIF-HKSLRPGUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Maduramicin ammonium belongs to the class of antibiotics employed as veterinary drugs to improve animal health and promote growth.

Application

Maduramicin ammonium may be used as an analytical reference standard for the quantification of the analyte in feeding stuffs using liquid chromatography coupled to tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohsin Raza et al.
Nanoscale, 10(28), 13773-13791 (2018-07-12)
Malaria continues to be one of the deadliest infectious diseases and a global health menace. The emergence and spread of drug-resistant strains of malaria parasites have further made the process of disease management grimmer. Thus, there is an urgent need
Ahmed Thabet et al.
Veterinary parasitology, 233, 86-96 (2017-01-04)
Availability of an accurate in vitro assay is a crucial demand to determine sensitivity of Eimeria spp. field strains toward anticoccidials routinely. In this study we tested in vitro models of Eimeria tenella using various polyether ionophores (monensin, salinomycin, maduramicin
Development of a multi-class method for the quantification of veterinary drug residues in feedingstuffs by liquid chromatography-tandem mass spectrometry
Boscher A, et al.
Journal of Chromatography A, 1217(41), 6394-6404 (2010)
Konrad Pietruk et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 35(8), 1508-1518 (2018-04-13)
A confirmatory multi-residue method was developed for the determination in milk of 19 coccidiostats (amprolium, arprinocid, clazuril, clopidol, decoquinate, diclazuril, ethopabate, halofuginone, lasalocid, maduramicin, monensin, narasin, nicarbazin, nequinate, robenidine, salinomycin, semduramicin, toltrazuril sulfone and toltrazuril sulfoxide). Sample preparation utilising extraction
Bartosz Sell et al.
Journal of analytical toxicology, 42(3), 183-191 (2017-12-02)
Reported here is a new analytical multiclass method based on QuEChERS technique, which has proven to be effective in diagnosing fatal poisoning cases in animals. This method has been developed for the determination of analytes in liver samples comprising rodenticides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service