Skip to Content
Merck
All Photos(1)

Key Documents

33431

Supelco

Iprovalicarb

PESTANAL®, analytical standard, mixture of isomers

Synonym(s):

Nα-(Isopropoxycarbonyl)-N-[1-(p-tolyl)ethyl]-L-valinamide, Isopropyl (S)-2-methyl-1-[1-(p-tolyl)ethylcarbamoyl]propylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28N2O3
CAS Number:
Molecular Weight:
320.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C1=CC=C(C=C1)C)NC([C@H](C(C)C)NC(OC(C)C)=O)=O

InChI

1S/C18H28N2O3/c1-11(2)16(20-18(22)23-12(3)4)17(21)19-14(6)15-9-7-13(5)8-10-15/h7-12,14,16H,1-6H3,(H,19,21)(H,20,22)/t14?,16-/m1/s1

InChI key

NWUWYYSKZYIQAE-BZSJEYESSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iprovalicarb belongs to the group of carboxylic acid amide fungicides with protective, curative, and eradicative systemic properties against plant pathogens such as Bremia lactucae, Plasmopara viticola and Phytophthora spp.

Application

Iprovalicarb may be used as a reference standard for the determination of iprovalicarb residues in agricultural and environmental samples using enzyme linked immunosorbent assay technique (ELISA).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baseline sensitivity and resistance-risk assessment of Phytophthora capsici to iprovalicarb.
Lu HX, et al.
Phytopathology, 100(11), 1162-1168 (2010)
Development of an ELISA for the detection of the residues of the fungicide iprovalicarb.
Lee KJ, et al.
Journal of Agricultural and Food Chemistry, 52(22), 6680-6686 (2004)
Noelia Briz-Cid et al.
International journal of molecular sciences, 20(6) (2019-03-27)
The effect of fungicides on fermentation is of paramount importance to control the quality and safety of wines. In this work, the quality (enological parameters, color, phenolic content, antioxidant activity, and fungicide residues) of wines from Monastrell grapes fortified with
Yengkhom Bijen Kumar et al.
Journal of food science and technology, 57(6), 2061-2069 (2020-05-21)
Dissipation behavior and degradation kinetics of fenamidone + mancozeb (Sectin 60 WG) and iprovalicarb + propineb (Melody Duo 66.75 WP) in tomato were studied at recommended dose (RD) and double dose (DD) of application. The analysis of the field samples were carried out by
Alice Agarbati et al.
PloS one, 14(6), e0217385-e0217385 (2019-06-21)
Yeast species that colonize the surface of grape berries at harvest time play an important role during the winemaking process. In this study, the use of culturable microbial techniques permitted a quantitative and qualitative inventory of the different yeast species

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service