Skip to Content
Merck
All Photos(1)

Key Documents

31593

Supelco

Arecoline hydrobromide

VETRANAL®, analytical standard

Synonym(s):

1-Methyl-1,2,5,6-tetrahydro-3-pyridinecarboxylic acid methyl ester hydrobromide, Arecaidine methyl ester hydrobromide, Methyl 1,2,5,6-tetrahydro-1-methyl-3-pyridinecarboxylate hydrobromide, Methyl 1-methyl-1,2,5,6-tetrahydronicotinate hydrobromide, Taeniolin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13NO2 · HBr
CAS Number:
Molecular Weight:
236.11
Beilstein:
3914826
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

172-176 °C (lit.)

application(s)

clinical
food and beverages

format

neat

SMILES string

Br[H].COC(=O)C1=CCCN(C)C1

InChI

1S/C8H13NO2.BrH/c1-9-5-3-4-7(6-9)8(10)11-2;/h4H,3,5-6H2,1-2H3;1H

InChI key

AXOJRQLKMVSHHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Arecoline hydrobromide is an arecoline salt of hydrobromic acid generally used in removing parasites from dog′s intestine. It has a strong parasympathomimetic action.

Application

Arecoline hydrobromide may have been used as working standard in the determination of arecoline concentration from dog plasma using LC–MS/MS.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah L Withey et al.
The Journal of pharmacology and experimental therapeutics, 366(2), 397-409 (2018-05-23)
Evidence suggests that the α4β2, but not the α7, subtype of the nicotinic acetylcholine receptor (nAChR) plays a key role in mediating the behavioral effects of nicotine and related drugs. However, the importance of other nAChR subtypes remains unclear. The
A Dakkak et al.
Acta tropica, 165, 26-32 (2016-07-21)
This study was undertaken in the Province of Sidi Kacem in northwest Morocco between April 2010 and March 2011. The main objective of the study was to determine the prevalence of Echinococcus granulosus (Eg) infection in owned dogs. This province
Sid Ahmed Labed et al.
Immunity, 48(5), 963-978 (2018-05-17)
Regulated antimicrobial peptide expression in the intestinal epithelium is key to defense against infection and to microbiota homeostasis. Understanding the mechanisms that regulate such expression is necessary for understanding immune homeostasis and inflammatory disease and for developing safe and effective
Yi-Ping Huang et al.
Clinical and experimental pharmacology & physiology, 44(8), 880-887 (2017-04-30)
Activation of peroxisome proliferator-activated receptor alpha (PPARα) has been reported to disrupt tumour metabolism and to promote anticancer activity through interfering with the Warburg effect. This study is to investigate whether Warburg effect-related proteins also could be identified in oral
Zhizhong Xu et al.
Neuroreport, 30(2), 134-138 (2018-12-21)
Cerebral demyelination is possibly one of the main pathological factors involved in the development of schizophrenia. Our previous studies have showed that Areca catechu nut extract could ameliorate cognitive decline by facilitating myelination processes in the frontal cortex in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service