Skip to Content
Merck
All Photos(2)

Key Documents

229148

Sigma-Aldrich

Purpurin

Dye content 90 %

Synonym(s):

1,2,4-Trihydroxyanthraquinone, Hydroxylizaric acid, Verantin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8O5
CAS Number:
Molecular Weight:
256.21
Colour Index Number:
58205
Beilstein:
1887127
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

>70-85% (HPLC)

form

powder

composition

Dye content, 90%

impurities

1-3% DMF

mp

253-256 °C (lit.)

solubility

NH4OH: 1 mg/mL, clear to turbid

λmax

515 nm
521 nm (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1cc(O)c2C(=O)c3ccccc3C(=O)c2c1O

InChI

1S/C14H8O5/c15-8-5-9(16)14(19)11-10(8)12(17)6-3-1-2-4-7(6)13(11)18/h1-5,15-16,19H

InChI key

BBNQQADTFFCFGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Purpurin (1, 2, 4-trihydroxyanthraquinone) is the key pigment present in the roots of Indian madder (Rubia cordifolia). This pigment produces colors, with distinctive heat and light resistant properties.

Application

Nuclear stain in histology

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of microwave-assisted extraction for alizarin and purpurin in Rubiaceae plants and its comparison with conventional extraction methods.
Dabiri M
Journal of Separation Science, 28, 387-396 (2005)
Dyeing studies with hydroxyanthraquinones extracted from Indian madder. Part 1: Dyeing of nylon with purpurin
Gupta D
Coloration Technology (2001)
C J Gomer et al.
Cancer research, 56(10), 2355-2360 (1996-05-15)
Photodynamic therapy (PDT) is an experimental cancer therapy inducing tumor tissue damage via photosensitizer-mediated oxidative cytotoxicity. A previous report indicates that oxidative stress induced by hydrogen peroxide or menadione activates the heat shock transcription factor in mouse cells but does
Modified porphyrins, chlorins, phthalocyanines, and purpurins: second-generation photosensitizers for photodynamic therapy.
M Kreimer-Birnbaum
Seminars in hematology, 26(2), 157-173 (1989-04-01)
Tim Marczylo et al.
Journal of agricultural and food chemistry, 51(11), 3334-3337 (2003-05-15)
Purpurin, an anthraquinone constituent from madder root, has previously been reported as antimutagenic in the Ames Salmonella bacterial mutagenicity assay and as antigenotoxic in Drosophila melanogaster, against a range of environmental carcinogens. Short-term dietary supplementation with purpurin inhibits the formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service