Skip to Content
Merck
All Photos(1)

Key Documents

10109878001

Roche

Trypsin Inhibitor

from chicken egg white

Synonym(s):

trypisin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

chicken egg white

Quality Level

form

powder

packaging

pkg of 1 g

manufacturer/tradename

Roche

concentration

1 mg/mL

technique(s)

cell culture | mammalian: suitable

optimum pH

3.8-4.5

solubility

water: soluble

storage temp.

2-8°C

General description

Egg white trypsin inhibitor is an inhibitor for trypsin, plasmin and plasma kallikrein activity in serum-free cell-culture media. It does not inhibit metallo-, cysteine, and aspartic proteases or tissue kallikrein (serine protease).

Application

Chicken egg Trypsin Inhibitor is used for the termination of tissue disaggregation and for subcultivation procedures.
Trypsin Inhibitor has been used:
  • as a component of lysis buffer to treat Tsc2fl/fl (control) and Tsc2fl/flLyz2-Cre (knockout [KO]) bone marrow-derived macrophages (BMDMs) before immunoblotting
  • to inactivate trypsin post detachment of the stem cells in ventricular zone cell culture
  • to neutralize trypsin post dissociation and trypsinization of brain tissues
  • termination of tissue disaggregation and for subcultivation procedures

Preparation Note

Working concentration: 0
Working solution: Solvent is recommended in distilled water.
Stock solution: 1 mg/ml
Storage conditions (working solution): -15 to -25 °C
Stable for at least 6 months in distilled water or 1 mM HCI when stored frozen in aliquots. Avoid repeated freeze-thaw cycles.

Reconstitution

Both are soluble in water. Recommended stock solution: 1 mg/ml. Store frozen in aliquots at -15 to -25 °C. Stable for at least 6 months.

Storage and Stability

Store at 2 to 8 °C. (Store dry!)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Membrane properties and electrogenesis in the distal axons of small dorsal root ganglion neurons in vitro.
Vasylyev DV and Waxman SG
Journal of Neurophysiology, 108(3), 729-740 (2012)
Maksim Sinyuk et al.
Journal of visualized experiments : JoVE, (160) (2020-06-23)
The methods presented here demonstrate laboratory procedures for the dissection of four different regions of the central nervous system (CNS) from murine neonates for the isolation of glial subpopulations. The purpose of the procedure is to dissociate microglia, oligodendrocyte progenitor
Haley M Peckham et al.
Journal of visualized experiments : JoVE, 95(95), 52179-52179 (2015-02-05)
Myelination is a complex process that involves both neurons and the myelin forming glial cells, oligodendrocytes in the central nervous system (CNS) and Schwann cells in the peripheral nervous system (PNS). We use an in vitro myelination assay, an established
Numb regulates Tau levels and prevents neurodegeneration in tauopathy mouse models.
Lacomme, et al.
Science Advances, 8, eabm4295-eabm4295 (2023)
June Bryan de la Peña et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 43(16), 2921-2933 (2023-03-10)
RNA stability is meticulously controlled. Here, we sought to determine whether an essential post-transcriptional regulatory mechanism plays a role in pain. Nonsense-mediated decay (NMD) safeguards against translation of mRNAs that harbor premature termination codons and controls the stability of ∼10%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service