Skip to Content
Merck
All Photos(4)

Key Documents

MAB1522

Sigma-Aldrich

Anti-Actin Antibody, smooth muscle γ & α actin, clone CGA7

ascites fluid, clone CGA7, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

clone

CGA7, monoclonal

species reactivity

human, rat, chicken, rabbit, monkey

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Antibody recognizes actin isotypes alpha and gamma of smooth muscle cells, but not skeletal or cardiac muscle cells. Reactive with myoepithelium, but nonreactive with other mesenchymal and all epithelial cells. The antibody labels eiomyosarcomas and leiomyomas but does not react with other neoplasms such as carcinomas or lymphoma.

Immunogen

Chicken gizzard actin
Epitope: smooth muscle gamma & alpha actin

Application

Detect Actin using this Anti-Actin Antibody, smooth muscle γ & α actin, clone CGA7 validated for use in WB, IH, IH(P).
Western Blot: 1:1000 (approximately 43 kDa)

Immunohistochemistry (paraffin, formalin, deparaffinized methacarn-fixed tissue: 1:10,000 or alcohol fixation)

Optimal dilutions must be determined by the end user.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional expression of the GABAB receptor in human airway smooth muscle.
Osawa, Y; Xu, D; Sternberg, D; Sonett, JR; D'Armiento, J; Panettieri, RA; Emala, CW
American Journal of Physiology. Lung Cellular and Molecular Physiology null
Cultured circular smooth muscle from the rabbit colon.
Kao, H W, et al.
In Vitro Cellular & Developmental Biology, 24, 787-794 (1988)
Liang Chen et al.
Journal of cardiovascular pharmacology, 68(3), 215-222 (2016-04-26)
The cardiotonic pill (CP), consisting of a mixture of Radix Salviae Miltiorrhizae, Radix Notoginseng, and Borneolum Syntheticum, has been widely used in the prevention and treatment of cardiovascular disease. Adhesion molecules, including intercellular cell adhesion molecule-1 and vascular cell adhesion
Xin Yao et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 13(1), 161-169 (2007-01-04)
Intratumoral microvascular density (MVD) has been controversial as an indicator of prognosis in clear cell renal cell carcinoma (CCRCC). Classification of the intratumoral blood vessels based on differential expressions of blood vessel markers has not been correlated with patient prognosis
Receptor-operated Ca2+ entry mediated by TRPC3/TRPC6 proteins in rat prostate smooth muscle (PS1) cell line.
S Thebault, A Zholos, A Enfissi, C Slomianny, E Dewailly, M Roudbaraki, J Parys, Natalia Prevarskaya
Journal of Cellular Physiology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service