Skip to Content
Merck
All Photos(3)

Key Documents

EZFITEXTE1

Millipore

EZ-Fit® Manifold Filtration Head Extension

For use with EZ-Fit® Manifold, 1 units

Synonym(s):

EZ-Fit® Filtration Manifold Accessory, EZ-Fit® Manifold

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

stainless steel

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 1 unit

manufacturer/tradename

EZ-Fit®

input

sample type bottled water
sample type beer
sample type cosmetics
sample type pharmaceutical(s)

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Height extension for EZ-FIT® Manifold is a part of the membrane filtration system and is made of stainless steel. It is an extension accessory designed specifically for operators working at benches. The package includes 1 unit of the height extension manifold.

Application

Height extension for EZ-FIT® Manifold is an accessory of the EZ-FIT® vacuum manifolds used during filtration of the test samples for general microbiological analysis, bioburden testing and water monitoring.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service