Skip to Content
Merck
All Photos(1)

Key Documents

317640-M

Sigma-Aldrich

Dipeptidyl Peptidase IV (CD26), Porcine Kidney

Synonym(s):

CD26, DPPIV

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥35 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−70°C

General description

Native, DPP IV purified from porcine kidney (renal brush border of the proximal tubule). A serine exopeptidase dimer composed of two identical subunits of 110-130 kDa. Cleaves Xaa-Pro dipeptides from the N-terminus of oligo and polypeptides. DPPIV is involved in many cellular processes such as activation of cytokines, differentiation, T cell activation, and cell-matrix interactions. Inhibition of DPPIV has been reported to be an effective treatment for type II diabetes. MW: ~210000-260000 (unreduced).

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmole 7-(Gly-Pro)-amino-4-methylcoumarinamide per min at 37°C, pH 8.5.

Physical form

In 20 mM Tris-HCl, 5 mM CaCl₂, 1 µM ZnCl₂, 0.05% NaN₃, pH 8.0.

Reconstitution

Dilute only the amount of enzyme needed for each assay. Upon thawing, do not re-freeze, store in the refrigerator (4°C).

Other Notes

Bar, J., et al. 2003. Biol. Chem.384, 1553.
Engel, M., et al. 2003. Proc. Natl. Acad. Sci. USA100, 5063.
Ikehara, Y., et al. 1994. Methods. Enzymol.244, 215.
David, F., et al. 1993. J. Biol. Chem.268, 17247.
Thomsen, P.D., et al. 1993. Mamm. Genome4, 604.
Misumi, Y., et al. 1992. Biochim. Biophys. Acta.1131, 333.
Seidl, R., et al. 1991. Biol. Chem. Hoppe Seyler372, 213.
Checler, F., et al. 1985. J. Neurochem.45, 1509.
Imai, K., et al. 1983. J. Biochem.93, 431.
Kato, T., et al. 1979. Experientia.35, 409.
Kojima, K., et al. 1979. Anal. Biochem.100, 43.
Kato, T., et al. 1978. Biochem. Med.19, 351.
Kenny, A.J., et al. 1976. Biochem. J.157, 169.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service