Skip to Content
Merck
All Photos(2)

Key Documents

1.05406

Millipore

Baird-Parker Agar

GranuCult® prime, ISO 6888, ISO 22178, FDA-BAM, APHA, granular, pack of 500 g

Synonym(s):

GranuCult® Baird-Parker agar (base), Staphylococcus selective agar, Staphylococcus selective agar acc. to BAIRD-PARKER (base)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

APHA
FDA-BAM
ISO 22178
ISO 6888

Quality Level

sterility

non-sterile

form

granular

packaging

pack of 500 g

manufacturer/tradename

GranuCult® prime

technique(s)

microbe id | specific enzyme detection: suitable
microbiological culture: suitable

pH

6.8 (37 °C, 58 g/L in H2O)

solubility

58 g/L

bulk density

570 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

Staphylococcus spp.

Looking for similar products? Visit Product Comparison Guide

General description

GranuCult® Baird-Parker agar (base) is used for the isolation and enumeration of Staphylococcus aureus. This medium contains lithium chloride and tellurite to inhibit the growth of accompanying microbial flora, whereas pyruvate and glycine selectively stimulate the growth of staphylococci. Agar is the solidifying agent. The egg-yolk content makes the medium opaque. Staphylococcus colonies show two characteristic features when grown in this medium: (a) characteristic zones and rings are formed as a result of lipolysis and proteolysis, and (b) reduction of tellurite to tellurium produces a black coloration. The egg-yolk reaction and tellurite reduction usually occur together with a positive coagulase reaction and thus can serve as an index for the latter. According to EN ISO 6888-1, the addition of sulfamethazine is advised to suppress the growth and swarming of Proteus sp., if these are suspected in the test sample.

Application

GranuCult® Baird-Parker agar (base) is used for the isolation, presumptive identification, and enumeration of Staphylococcus aureus from food and animal feed as well as from other materials.

Linkage

Replaces: 1.05406.5007

Analysis Note

Appearance (clearness): clear to turbid
Appearance (color): yellowish-brown
pH-value (25 °C): 6.6 - 7.0
Solidification behaviour (2 hrs., 45 °C): liquid
Growth promotion test in accordance with the current version of DIN EN ISO 11133.
Inoculum on reference medium (Staphylococcus aureus ATCC 6538 (WDCM 00032)):
Inoculum on reference medium (Staphylococcus aureus ATCC 25923 (WDCM 00034)):
Colony count (Staphylococcus aureus ATCC 6538 (WDCM 00032)):
Colony count (Staphylococcus aureus ATCC 25923 (WDCM 00034)):
Recovery on test medium (Staphylococcus aureus ATCC 6538 (WDCM 00032)): ≥ 50 %
Recovery on test medium (Staphylococcus aureus ATCC 25923 (WDCM 00034)): ≥ 50 %
Growth (Escherichia coli ATCC 8739 (WDCM 00012)): no growth
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): no growth
Growth (Staphylococcus saprophyticus ATCC 15305 (WDCM 00159)): no limit
Growth (Staphylococcus epidermidis ATCC 12228 (WDCM 00036)): no limit
Growth (Enterococcus hirae ATCC 8043 (WDCM 00089)): no limit
Growth (Proteus mirabilis ATCC 29906 (WDCM 00023)): no limit
Colonies (Staphylococcus aureus ATCC 6538 (WDCM 00032)): Black or grey colonies with clear halo
Colonies (Staphylococcus aureus ATCC 25923 (WDCM 00034)): Black or grey colonies with clear halo
Colonies (Staphylococcus saprophyticus ATCC 15305 (WDCM 00159)): Black or grey colonies without clear halo
Colonies (Staphylococcus epidermidis ATCC 12228 (WDCM 00036)): Black or grey colonies without clear halo
Colonies (Enterococcus hirae ATCC 8043 (WDCM 00086)): Black or grey colonies without clear halo
Colonies (Proteus mirabilis ATCC 29906 (WDCM 00023)): Brown or black colonies without clear halo
Incubation:
24 ± 2 to 48 ± 2 hours at 37 ± 1 °C aerobic
E.coli 48 ± 2 hours at 37 ± 1 °C aerobic
A recovery rate of 50 % is equivalent to a productivity value of 0.5.
The indicated colony counts result from the sum of a triple determination.
Reference media: Tryptic Soy Agar

Other Notes

The prepared medium is clear to turbid and yellowish-brown.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Tenderis et al.
International journal of food microbiology, 321, 108560-108560 (2020-02-23)
Foodborne illnesses affect the health of consumers worldwide, and thus searching for potential antimicrobial agents against foodborne pathogens is given an increased focus. This research evaluated the influence of sodium lactate (SL), encapsulated (e) and unencapsulated (u) polyphosphates (PP; sodium

Articles

Microbial culture media is available in both powdered and granulated forms. This article compares the characteristics of each culture media format with regards to safety, handling and convenience.

Microbial culture media is available in both powdered and granulated forms. This article compares the characteristics of each culture media format with regards to safety, handling and convenience.

Microbial culture media is available in both powdered and granulated forms. This article compares the characteristics of each culture media format with regards to safety, handling and convenience.

Microbial culture media is available in both powdered and granulated forms. This article compares the characteristics of each culture media format with regards to safety, handling and convenience.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service