Skip to Content
Merck
All Photos(1)

Key Documents

R-014

Supelco

(±)-threo-Ritalinic acid-d10 hydrochloride

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(±)-threo-Ritalinic acid-d10 HCl solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H7NO2D10
Molecular Weight:
229.34
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.[2H]C1([2H])N[C@@]([2H])(C([2H])([2H])C([2H])([2H])C1([2H])[2H])[C@]([2H])(C(O)=O)c2ccccc2

InChI

1S/C13H17NO2.ClH/c15-13(16)12(10-6-2-1-3-7-10)11-8-4-5-9-14-11;/h1-3,6-7,11-12,14H,4-5,8-9H2,(H,15,16);1H/t11-,12+;/m0./s1/i4D2,5D2,8D2,9D2,11D,12D;

InChI key

SCUMDQFFZZGUQY-SMKJLJITSA-N

General description

Ritalinic acid is the major urinary metabolite of methylphenidate, more commonly known by the brand name Ritalin® which is approved for treatment of ADHD (Attention-Deficit Hyperactivity Disorder) and narcolepsy. This new stable-labeled internal standard is suitable for a wide variety of GC/MS and LC/MS applications from clinical toxicology and urine drug testing to forensic analysis and isotope dilution methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Ritalin is a registered trademark of Novartis Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service