Skip to Content
Merck
All Photos(1)

Key Documents

P-096

Supelco

Pregabalin-D6 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8D6H11NO2
Molecular Weight:
165.26
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1=C2C(NC(CCOCCC(O)=O)=O)=CC=C1)N([C@H]3CCC(NC3=O)=O)C2=O.O=C(C4=C5C(NC(CCOCCC(O)=O)=O)=CC=C4)N([C@H]6CCC(NC6=O)=O)C5=O

InChI

1S/C8H17NO2/c1-6(2)3-7(5-9)4-8(10)11/h6-7H,3-5,9H2,1-2H3,(H,10,11)/i1D3,2D3

InChI key

AYXYPKUFHZROOJ-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sold under the trade name Lyrica®, Pregabalin is an antiepileptic drug used for treatment of seizures, fibromyalgia, diabetic peripheral neuropathy, generalized anxiety disorder, and chronic neuropathic pain. This internal standard is suitable for pregabalin GC/MS or LC/MS testing or isotope dilution methods in forensic analysis, clinical toxicology, pain prescription monitoring, or urine drug testing.

Application


  • Pregabalin-D₆ in Therapeutic Drug Monitoring: Pregabalin-D₆ serves as a crucial analytical standard in therapeutic drug monitoring using mass spectrometry, enhancing the precision of quantifying new-generation antiseizure medications among epilepsy patients (Kim et al., 2024).

  • Multi-Component LC-MS/MS Analysis: Utilizing Pregabalin-D₆ as an internal standard, a multicomponent LC-MS/MS method was developed for drugs of abuse testing, showcasing its utility in comprehensive bioanalytical assays (Guterstam et al., 2024).

  • Comprehensive Drug Class Screening: The use of Pregabalin-D₆ in the determination of various drug classes in whole blood exemplifies its role in complex toxicological screenings using LC-MS/MS, facilitating accurate metabolic studies (Jørgenrud et al., 2021).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Lyrica is a registered trademark of C.P. Pharmaceuticals International C.V.
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service