Skip to Content
Merck
All Photos(1)

Key Documents

890899C

Avanti

DODMA

Avanti Research - A Croda Brand

Synonym(s):

1,2-Dioleyloxy-3-dimethylaminopropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H81NO2
CAS Number:
Molecular Weight:
620.09
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 4 mL (890899C-100MG)
pkg of 5 × 4 mL (890899C-500MG)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

25 mg/mL in chloroform

application(s)

advanced drug delivery

lipid type

cationic lipids
transfection

shipped in

dry ice

storage temp.

−20°C

SMILES string

CN(C)CC(OCCCCCCCC/C=C\CCCCCCCC)COCCCCCCCC/C=C\CCCCCCCC

InChI

1S/C41H81NO2/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-43-40-41(39-42(3)4)44-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2/h19-22,41H,5-18,23-40H2,1-4H3

InChI key

GLGLUQVVDHRLQK-UHFFFAOYSA-N

General description

DODMA (1,2-dioleyloxy-3-dimethylaminopropane) is an ionizable cationic lipid. DODMA possesses one double bond in each hydrocarbon tail with tertiary amine group and has a pKa of 7.0.

Application

DODMA is suitable for use in the preparation of lipid nanoparticles (LNPs).

Biochem/physiol Actions

DODMA (1,2-dioleyloxy-3-dimethylaminopropane) is used in the preparation of cationic liposomes and lipid nanoparticles (LNPs). LNPs with DODMA has low or neutral zeta potential. DODMA is efficient in encapsulating nucleic acid during synthesis by reducing the physiological pH. DODMA is implicated in gene therapy.

Packaging

5 mL Clear Glass Sealed Ampule (890899C-100MG)
5 × 5 mL Clear Glass Sealed Ampule (890899C-500MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of the nanoformulation of siRNA-lipid assemblies on their cellular uptake and immune stimulation
Kubota K, et al.
International journal of nanomedicine, 12(3), 5121-5121 (2017)
Lipofection of plasmid DNA into human mast cell lines using lipid nanoparticles generated by microfluidic mixing
Duguay BA, et al.
Journal of Leukocyte Biology, 104(3), 587-596 (2018)
Application of DODMA and derivatives in cationic nanocarriers for gene delivery
Chen Z, et al.
Current Organic Chemistry, 20(17), 1813-1819 (2016)
Lipid nanoparticles for delivery of messenger RNA to the back of the eye
Patel S, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 303(3), 91-100 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service