Key Documents
Z677477
Camatril Velours nitrile flock lined gloves
size XXL
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
green gloves
green nitrile
size
XXL
sterility
non-sterile
feature
CE compliant
disposable
latex free
powder-free
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
packaging
package of 100 ea
manufacturer/tradename
KCL 730 - 11
KCL 730
L × thickness
300 mm × 0.4 mm
General description
Camatril Velours green nitrile flock lined gloves are manufactured from high quality nitrile ensuring effective chemical resistance for use in numerous fields. The remarkably high abrasion, cut and puncture resistance allows work with sharp edged materials.
Good flexibility, anatomical design and excellent fit ensure a good sense of touch. The flock lining offers additional wearing comfort.
  • Ergonomic fit ensures good sensitivity
  • Roughened palm for excellent grip when wet
  • High mechanical resistance
  • Cotton flock lining improves comfort
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • CE compliant
Legal Information
Camatril is a trademark of KCL GmbH