Skip to Content
Merck
All Photos(1)

Key Documents

721905

Sigma-Aldrich

Gadolinium(III) chloride solution

NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), Methanol-13C 0.1 % (99 atom % 13C), water 1 %, NMR tube size 3 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GdCl3
CAS Number:
Molecular Weight:
263.61
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.12

grade

NMR reference standard

form

solution

composition

Methanol-13C, 0.1% (99 atom % 13C)
water, 1%

concentration

0.1 mg/mL in D2O (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

3 mm × 8 in.

SMILES string

Cl[Gd](Cl)Cl

InChI

1S/3ClH.Gd/h3*1H;/q;;;+3/p-3

InChI key

MEANOSLIBWSCIT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Auto test

Quantity

3 mm O.D. tube contains 0.235 mL.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianjing Li et al.
Immunologic research, 62(2), 137-145 (2015-04-08)
In this study, we found that pretreatment with low dose of lipopolysaccharide (LPS), also known as lipoglycans and endotoxin, obviously attenuated liver injury caused by diethylnitrosamine (DEN) in mice. This protective effect was described by decreased ALT, TNF-α, and IL-1β
Xiaojuan Chai et al.
Nature communications, 6, 7979-7979 (2015-08-11)
Sepsis is the host's deleterious systemic inflammatory response to microbial infections. Here we report an essential role for the oestrogen sulfotransferase (EST or SULT1E1), a conjugating enzyme that sulfonates and deactivates estrogens, in sepsis response. Both the caecal ligation and
Quirin Zangl et al.
Anesthesiology, 121(6), 1217-1225 (2014-09-17)
Liver damage by ischemia and reperfusion injury is a risk factor for morbidity and mortality after liver surgery. Postoperative oxygen treatment is routinely applied in the postanesthesia and intensive care unit after liver surgery. The risks of aggravating the injury
Yan Yang et al.
European journal of immunology, 44(8), 2468-2477 (2014-04-30)
Basophils have been shown to contribute to anaphylaxis through either an IgE-FcεRI-dependent pathway or an IgG-FcγR pathway. However, it remains largely unclear whether basophils can be activated to promote anaphylaxis via a non-FcR pathway as well. The glycolipid receptor ASGM1
Hiroaki Kito et al.
Biochemical and biophysical research communications, 459(3), 457-462 (2015-03-10)
Store-operated Ca(2+) entry (SOCE) via Orai1 and STIM1 complex is supposed to have obligatory roles in the regulation of cellular functions of vascular endothelial cells, while little is known about the contribution of Orai2. Quantitative PCR and Western blot analyses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service