Skip to Content
Merck
All Photos(1)

Key Documents

485799

Sigma-Aldrich

Hexadecane-1,2-13C2

99 atom % 13C

Synonym(s):

13C Labeled hexadecane, Cetane-1,2-13C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)1313CH213CH3
CAS Number:
Molecular Weight:
228.43
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

refractive index

n20/D 1.434 (lit.)

bp

287 °C (lit.)

mp

18 °C (lit.)

density

0.780 g/mL at 25 °C

mass shift

M+2

SMILES string

CCCCCCCCCCCCCC[13CH2][13CH3]

InChI

1S/C16H34/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3-16H2,1-2H3/i1+1,3+1

InChI key

DCAYPVUWAIABOU-ZKDXJZICSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

275.0 °F - closed cup

Flash Point(C)

135 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service