Skip to Content
Merck
All Photos(1)

Key Documents

485365

Sigma-Aldrich

Caffeine-(trimethyl-13C3)

99 atom % 13C, 99% (CP)

Synonym(s):

1H-Purine-2,6-dione, 3,7-dihydro-1,3,7-tri(methyl-13C)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C5H10N4O2
CAS Number:
Molecular Weight:
197.17
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

234-236.5 °C (lit.)

mass shift

M+3

SMILES string

[13CH3]N1C(=O)N([13CH3])c2ncn([13CH3])c2C1=O

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i1+1,2+1,3+1

InChI key

RYYVLZVUVIJVGH-VMIGTVKRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Caffeine-(trimethyl-13C3) is an isotopic analogue of L-caffeine.

Application

L-Caffeine-(trimethyl-13C3) can be used
in the determination of L-Ephedrine, pseudooephedrine and caffeine in rat plasma using LC-TMS and study its toxicokinetic value.
as an internal standard for the investigation of caffeine concentration in drinking foods.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeroen Peters et al.
Analytical and bioanalytical chemistry, 405(24), 7783-7794 (2013-06-14)
A combined (triplex) immunoassay for the simultaneous detection of three mycotoxins in grains was developed with superparamagnetic colour-encoded microbeads, in combination with two bead-dedicated flow cytometers. Monoclonal antibodies were coupled to the beads, and the amounts of bound mycotoxins were
Stephen D Cooper et al.
Journal of analytical toxicology, 35(6), 341-348 (2011-07-12)
A rapid and simple liquid chromatography tandem mass spectrometry method was developed and validated for the simultaneous determination of L-ephedrine, pseudoephedrine, and caffeine in male Fisher-344 rat plasma at nanogram-per-milliliter concentrations for use in support of toxicology studies. Only 25
Noelia López-Gutiérrez et al.
Food chemistry, 173, 607-618 (2014-12-04)
A method has been developed and validated for the simultaneous detection and quantification of phytochemicals in nutraceutical products obtained from green tea. For that purpose, ultra-high performance liquid chromatography coupled to single-stage Orbitrap high resolution mass spectrometry (UHPLC-Orbitrap-MS) has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service