Skip to Content
Merck
All Photos(1)

Key Documents

476552

Sigma-Aldrich

Vinylmagnesium chloride solution

1.6 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHMgCl
CAS Number:
Molecular Weight:
86.80
Beilstein:
3535268
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

Quality Level

concentration

1.6 M in THF

density

0.978 g/mL at 25 °C

SMILES string

Cl[Mg]C=C

InChI

1S/C2H3.ClH.Mg/c1-2;;/h1H,2H2;1H;/q;;+1/p-1

InChI key

NAFZXKWEUWVSAW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used with iron pentadienoate complexes in a synthesis of divinyl cyclopropanes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 3639-3639 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service