Skip to Content
Merck
All Photos(1)

Key Documents

184667

Sigma-Aldrich

(Chloromethyl)cyclopropane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5CH2Cl
CAS Number:
Molecular Weight:
90.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.435 (lit.)

bp

87-89 °C (lit.)

density

0.98 g/mL at 25 °C (lit.)

SMILES string

ClCC1CC1

InChI

1S/C4H7Cl/c5-3-4-1-2-4/h4H,1-3H2

InChI key

ZVTQWXCKQTUVPY-UHFFFAOYSA-N

General description

(Chloromethyl)cyclopropane on reaction with isolable dialkylsilylene affords 5-silaspiro[4.4]nonane derivative.

Application

(Chloromethyl)cyclopropane was used in the synthesis of (lithiomethyl)cyclopropane and homoallyllithium via 4,4′-di-tert-butylbiphenyl or naphthalene-catalysed lithiation reaction.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unexpected reactions of an isolable dialkylsilylene with Haloalkanes.
Ishida S, et al.
Chemistry Letters (Jpn), 11, 1102-1103 (2001)
Easy selective generation of (lithiomethyl) cyclopropane or homoallyllithium by a chlorine-lithium exchange.
Pastor IM, et al.
Tetrahedron Letters, 49(48), 6870-6872 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service