Skip to Content
Merck
All Photos(2)

Key Documents

107271

Sigma-Aldrich

(±)-3-Chloro-1,2-propanediol

98%

Synonym(s):

α-Chlorohydrin, α-Glycerol chlorohydrin, α-Monochlorohydrin, 3-MCPD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
110.54
Beilstein:
635684
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.04 mmHg ( 25 °C)

Assay

98%

refractive index

n20/D 1.480 (lit.)

bp

213 °C (lit.)

solubility

H2O: soluble
alcohol: soluble
diethyl ether: soluble

density

1.322 g/mL at 25 °C (lit.)

SMILES string

OCC(O)CCl

InChI

1S/C3H7ClO2/c4-1-3(6)2-5/h3,5-6H,1-2H2

InChI key

SSZWWUDQMAHNAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Chloro-1,2-propanediol undergoes enantioselective transformation to (R)-3-chloro-1,2-propanediol by the cell extract of Corynebacterium sp. strain N-1074 . It is a contaminant found in food in free (diol) form as well as in an esterified (with fatty acids) form.

Application

(±)-3-Chloro-1,2-propanediol may be used in manufacture of dye intermediates.

Biochem/physiol Actions

3-Chloro-1,2-propanediol is a potent rodent chemosterilant.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Irrit. 2 - STOT RE 1 - STOT SE 1

Target Organs

Kidney, Testes

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Antifertility activity of 3-chloro-1, 2-propanediol (U-5897) on male rats.
E Samojlik et al.
Biology of reproduction, 2(2), 299-304 (1970-04-01)
T Nakamura et al.
Journal of bacteriology, 174(23), 7613-7619 (1992-12-01)
During the course of the transformation of 1,3-dichloro-2-propanol (DCP) into (R)-3-chloro-1,2-propanediol [(R)-MCP] with the cell extract of Corynebacterium sp. strain N-1074, epichlorohydrin (ECH) was transiently formed. The cell extract was fractionated into two DCP-dechlorinating activities (fractions Ia and Ib) and
W Seefelder et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 25(4), 391-400 (2008-03-19)
3-Mono-chloropropane-1,2-diol (3-MCPD) is a contaminant that occurs in food in its free (diol) form as well as in an esterified (with fatty acids) form. Using a simple intestinal model, it was demonstrated that 3-MCPD monoesters and 3-MCPD diesters are accepted
Ching-Yu Lin et al.
Reproductive toxicology (Elmsford, N.Y.), 28(1), 75-80 (2009-06-06)
Sperm ATP is derived primarily from either glycolysis or mitochondrial oxidative phosphorylation. In the present studies, (1)H NMR spectroscopy was used to characterize the metabolite profile in primate sperm treated either with alpha-chlorohydrin (ACH), a known inhibitor of sperm glycolysis
Z Zelinková et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 25(6), 669-676 (2008-05-20)
A series of twelve breast milk samples were analysed by gas chromatography-mass spectrometry (GC/MS) operated in selected ion monitoring mode for 3-chloropropane-1,2-diol (3-MCPD). Whilst none of the samples contained 3-MCPD above the limit of detection of 3 microg kg(-1) milk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service