HPLC Analysis of α-Methylbenzylamine Enantiomers (3,5-Dinitrobenzoyl Derivatives) on Astec® CHIROBIOTIC® V
Materials
analytical column
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] hexane; [B] isopropanol; (90:10, A:B)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
1 μL
sample
2 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC