Merck
A Unique ISR Program Determines Cellular Responses to Chronic Stress.
Molecular cell (2017-12-09)
Bo-Jhih Guan, Vincent van Hoef, Raul Jobava, Orna Elroy-Stein, Leos S Valasek, Marie Cargnello, Xing-Huang Gao, Dawid Krokowski, William C Merrick, Scot R Kimball, Anton A Komar, Antonis E Koromilas, Anthony Wynshaw-Boris, Ivan Topisirovic, Ola Larsson, Maria Hatzoglou
ABSTRACT
The integrated stress response (ISR) is a homeostatic mechanism induced by endoplasmic reticulum (ER) stress. In acute/transient ER stress, decreased global protein synthesis and increased uORF mRNA translation are followed by normalization of protein synthesis. Here, we report a dramatically different response during chronic ER stress. This chronic ISR program is characterized by persistently elevated uORF mRNA translation and concurrent gene expression reprogramming, which permits simultaneous stress sensing and proteostasis. The program includes PERK-dependent switching to an eIF3-dependent translation initiation mechanism, resulting in partial, but not complete, translational recovery, which, together with transcriptional reprogramming, selectively bolsters expression of proteins with ER functions. Coordination of transcriptional and translational reprogramming prevents ER dysfunction and inhibits "foamy cell" development, thus establishing a molecular basis for understanding human diseases associated with ER dysfunction.
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
MISSION® esiRNA, targeting human FTO
Sigma-Aldrich
MISSION® esiRNA, targeting human METTL3
Sigma-Aldrich
MISSION® esiRNA, targeting human METTL4
Sigma-Aldrich
ISRIB, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human PDCD4
Sigma-Aldrich
Cyclopiazonic acid from Penicillium cyclopium, ≥98% (HPLC), powder
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
N6-Cyclopentyladenosine, solid
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film