Merck
  • Atg9A trafficking through the recycling endosomes is required for autophagosome formation.
Atg9A trafficking through the recycling endosomes is required for autophagosome formation.
Journal of cell science (2016-09-03)
Kenta Imai, Feike Hao, Naonobu Fujita, Yasuhiro Tsuji, Yukako Oe, Yasuhiro Araki, Maho Hamasaki, Takeshi Noda, Tamotsu Yoshimori
ABSTRACT
Autophagy is an intracellular degradation pathway conserved in eukaryotes. Among core autophagy-related (Atg) proteins, mammalian Atg9A is the sole multi-spanning transmembrane protein, and both of its N- and C-terminal domains are exposed to the cytoplasm. It is known that Atg9A travels through the trans-Golgi network (TGN) and the endosomal system under nutrient-rich conditions, and transiently localizes to the autophagosome upon autophagy induction. However, the significance of Atg9A trafficking for autophagosome formation remains elusive. Here, we identified sorting motifs in the N-terminal cytosolic stretch of Atg9A that interact with the adaptor protein AP-2. Atg9A with mutations in the sorting motifs could not execute autophagy and was abnormally accumulated at the recycling endosomes. The combination of defects in autophagy and Atg9A accumulation in the recycling endosomes was also found upon the knockdown of TRAPPC8, a specific subunit of the TRAPPIII complex. These results show directly that the trafficking of Atg9A through the recycling endosomes is an essential step for autophagosome formation.
Product Description
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Anti-WIPI-2 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture