Merck
  • A potentiometric flow biosensor based on ammonia-oxidizing bacteria for the detection of toxicity in water.
A potentiometric flow biosensor based on ammonia-oxidizing bacteria for the detection of toxicity in water.
Sensors (Basel, Switzerland) (2013-05-28)
Qianyu Zhang, Jiawang Ding, Lijuan Kou, Wei Qin
ABSTRACT
A flow biosensor for the detection of toxicity in water using the ammonia-oxidizing bacterium (AOB) Nitrosomonas europaea as a bioreceptor and a polymeric membrane ammonium-selective electrode as a transducer is described. The system is based on the inhibition effects of toxicants on the activity of AOB, which can be evaluated by measuring the ammonium consumption rates with the ammonium-selective membrane electrode. The AOB cells are immobilized on polyethersulfone membranes packed in a holder, while the membrane electrode is placed downstream in the flow cell. Two specific inhibitors of the ammonia oxidation-allylthiourea and thioacetamide-have been tested. The IC50 values defined as the concentration of an inhibitor causing a 50% reduction in the ammonia oxidation activity have been measured as 0.17 μM and 0.46 μM for allylthiourea and thioacetamide, respectively. The proposed sensor offers advantages of simplicity, speed and high sensitivity for measuring toxicity in water.
Product Description
Supelco
Potassium tetrakis(4-chlorophenyl)borate, for ion-selective electrodes, Selectophore, ≥98.0%