Merck
  • Effect of the degree of substitution in the transition temperatures and hydrophobicity of hydroxypropyl cellulose esters.
Effect of the degree of substitution in the transition temperatures and hydrophobicity of hydroxypropyl cellulose esters.
Carbohydrate polymers (2015-04-11)
Delia López-Velázquez, Armando R Hernández-Sosa, Ernesto Pérez
ABSTRACT
We have synthesized and characterized five members of a homologous series of side chain polymers of hydroxypropyl cellulose esters obtained by homogeneous esterification with 6-[4'-(ethoxycarbonyl)biphenyl-4-yloxy]hexanoic acid. Two acylation procedures were studied. One procedure involved the acid chloride derivative and the other one was the activation of that acid with N,N'-carbonyldiimidazol. The second method yielded esters with higher degree of substitution, DS¯. The esters, with DS¯ ranging from 26 to about 66%, were characterized by FTIR, NMR, solution viscometry, TGA, DSC, polarized optical microscopy, and X-ray diffraction, in order to study the effect of the degree of substitution on the hydrophobicity, on the transition temperatures and on their potential liquid crystal properties. It has been found that the hydrophobicity and the transition temperatures of the HPC derivatives are very much dependent on the degree of acylation. The esters with high DS¯ values showed liquid crystal properties.
Sigma-Aldrich
CDI, ≥97.0% (T)
Sigma-Aldrich
Tetrabutylammonium fluoride on silica gel, ~1.5 mmol/g (F-)
Sigma-Aldrich
CDI, reagent grade
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 1.0 M in THF
Sigma-Aldrich
Tetrabutylammonium fluoride trihydrate, ≥97.0% (NT)
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 75 wt. % in H2O