Merck
  • Nanoparticle delivery of donor antigens for transplant tolerance in allogeneic islet transplantation.
Nanoparticle delivery of donor antigens for transplant tolerance in allogeneic islet transplantation.
Biomaterials (2014-07-30)
Jane Bryant, Kelan A Hlavaty, Xiaomin Zhang, Woon-Teck Yap, Lei Zhang, Lonnie D Shea, Xunrong Luo
ABSTRACT
Human islet cell transplantation is a promising treatment for type 1 diabetes; however, long-term donor-specific tolerance to islet allografts remains a clinically unmet goal. We have previously shown that recipient infusions of apoptotic donor splenocytes chemically treated with 1-ethyl-3-(3'-dimethylaminopropyl)-carbodiimide (donor ECDI-SP) can mediate long-term acceptance of full major histocompatibility complex (MHC)-mismatched murine islet allografts without the use of immunosuppression. In this report, we investigated the use of poly(lactide-co-glycolide) (PLG) particles in lieu of donor ECDI-SP as a synthetic, cell-free carrier for delivery of donor antigens for the induction of transplant tolerance in full MHC-mismatched murine allogeneic islet transplantation. Infusions of donor antigen-coupled PLG particles (PLG-dAg) mediated tolerance in ∼20% of recipient mice, and the distribution of cellular uptake of PLG-dAg within the spleen was similar to that of donor ECDI-SP. PLG-dAg mediated the contraction of indirectly activated T cells but did not modulate the direct pathway of allorecognition. Combination of PLG-dAg with a short course of low dose immunosuppressant rapamycin at the time of transplant significantly improved the tolerance efficacy to ∼60%. Furthermore, altering the timing of PLG-dAg administration to a schedule that is more feasible for clinical transplantation resulted in equal tolerance efficacy. Thus, the combination therapy of PLG-dAg infusions with peritransplant rapamycin represents a clinically attractive, biomaterials-based and cell-free method for inducing long-term donor-specific tolerance for allogeneic cell transplantation, such as for allogeneic islet transplantation.
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
2-Ethyl-2-phenylmalonamide monohydrate, 99%
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Supelco
Rapamycin, VETRANAL®, analytical standard