Merck
  • Dilution of semi-solid creams: influence of various production parameters on rheological properties and skin penetration.
Dilution of semi-solid creams: influence of various production parameters on rheological properties and skin penetration.
International journal of pharmaceutics (2014-12-06)
C Nagelreiter, E Kratochvilova, C Valenta
ABSTRACT
In order to customise treatment for patients, topical formulations are often diluted with drug-free cream bases to adjust the drug dose and thereby the formulations' activity to the patients' needs. However, the process of dilution influences properties of the formulations. Stability can be reduced as well as the microbial stability and most importantly, efficacy and skin penetration behaviour can be severely and unpredictably changed. The present study investigates the effects of production parameters on creams, namely incorporation of an API (active pharmaceutical ingredients) into an OW cream with prior mixing with propylene glycol or without and subsequent automated or manual dilution of the resulting creams with three different cream bases. Effects were measured by influence on microscopic appearance, measurement of chemical stability, skin penetration and rheological behaviour. suggest strong influence of the cream bases used for dilution of the formulations. Mixture of equal amounts of the employed OW and WO cream proved unfavourable due to inferior penetration behaviour and less appealing microscopic and macroscopic appearance. Prior mixing with PG was of negligible importance for the characteristics of the dilutions, however, the type of API and manner of dilution had an influence on the viscosity of the formulations.
Product Description
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Diclofenac sodium salt, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Fludrocortisone acetate, ≥98%
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Fludrocortisone acetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ammonium acetate, for inorganic trace analysis, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%