Merck
  • Clinical application of immunomagnetic reduction for quantitative measurement of insulin-like growth factor binding protein-1 in the prediction of pregnant women with preterm premature rupture of membranes.
Clinical application of immunomagnetic reduction for quantitative measurement of insulin-like growth factor binding protein-1 in the prediction of pregnant women with preterm premature rupture of membranes.
Clinica chimica acta; international journal of clinical chemistry (2014-10-05)
Chen-Yu Chen, Chia-Chen Chang, Chii-Wann Lin
ABSTRACT
Insulin-like growth factor binding protein-1 (IGFBP-1) constitutes a subgroup of the insulin-like growth factor binding protein systems, and its concentration in amniotic fluid is 100-1000 times higher than the concentration in other body fluids. The aim of this study was to evaluate the clinical application of a novel method immunomagnetic reduction (IMR) for quantitative measurement of IGFBP-1 concentrations in the cervicovaginal secretions to diagnose pregnant women with preterm premature rupture of membranes (PPROM). We established a standard calibration curve of IMR intensity against IGFBP-1 concentration based on standard IGFBP-1 samples. We used the IMR assay to detect IGFBP-1 concentrations in the cervicovaginal secretions of pregnant women which were divided into two groups according to the presence or absence of PPROM. The calibration curve extended from 0.1ng/mL to 10000ng/mL with an excellent correlation (R(2)=0.999). Twenty-two pregnant women between 22 and 34weeks of gestation were analyzed in this prospective study, of whom 10 were clinical evidence of PPROM, and 12 were intact membranes. Through the analysis of receiver-operating characteristic curve, the cut-off point for IMR to differentiate intact membranes from PPROM is 1.015%, which resulted in 90.0, 83.3, 81.8, and 90.9% for sensitivity, specificity, positive predictive value, and negative predictive value, respectively. It is evidenced that IMR assay can quantitatively analyze IGFBP-1 concentrations, and the results show the possibility to diagnose pregnant women with PPROM by IMR assay.
Sigma-Aldrich
Iron(III) chloride, sublimed grade, ≥99.9% trace metals basis
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Millipore
TDA Reagent, suitable for microbiology
Sigma-Aldrich
Iron(III) chloride solution, purum, 45% FeCl3 basis
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran