Merck
  • Anti-Eimeria activity of berberine and identification of associated gene expression changes in the mouse jejunum infected with Eimeria papillata.
Anti-Eimeria activity of berberine and identification of associated gene expression changes in the mouse jejunum infected with Eimeria papillata.
Parasitology research (2015-02-11)
Mohamed A Dkhil, Mahmoud S Metwaly, Saleh Al-Quraishy, Nour E Sherif, Denis Delic, Suliman Y Al Omar, Frank Wunderlich
ABSTRACT
Plant-based natural products are promising sources for identifying novel agents with potential anti-Eimeria activity. This study explores possible effects of berberine on Eimeria papillata infections in the jejunum of male Swiss albino mice. Berberine chloride, when daily administered to mice during infection, impairs intracellular development and multiplication of E. papillata, evidenced as 60% reduction of maximal fecal output of oocysts on day 5 p.i. Concomitantly, berberine attenuates the inflammatory response, evidenced as decreased messenger RNA (mRNA) expression of IL-1β, IL-6, TNFα, IFNγ, and iNOS, as well as the oxidative stress response, evidenced as impaired increase in malondialdehyde, nitrate, and H2O2 and as prevented decrease in glutathione and catalase activity. Berberine also alters gene expression in the infected jejunum. On day 5 p.i., mRNA expression of 29 genes with annotated functions is more than 10-fold upregulated and that of 14 genes downregulated. Berberine downregulates the genes Xaf1, Itgb3bp, and Faim3 involved in apoptotic processes and upregulates genes involved in innate immune responses, as e.g., Colec11, Saa2, Klra8, Clec1b, and Crtam, especially the genes Cpa3, Fcer1a, and Mcpt1, Mcpt2, and Mcpt4 involved in mast cell activity. Additionally, 18 noncoding lincRNA species are differentially expressed more than 10-fold under berberine. Our data suggest that berberine induces hosts to exert anti-Eimeria activity by attenuating the inflammatory and oxidative stress response, by impairing apoptotic processes, and by activating local innate immune responses and epigenetic mechanisms in the host jejunum. Berberine has the potential as an anti-Eimeria food additive in animal farming.
Sigma-Aldrich
Nitrogen, ≥99.998%
Berberine chloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Berberine chloride form
Supelco
Berberine Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Berberine chloride hydrate, technical, ≥90% (AT)