Merck
  • Identification of the effective constituents for anti-inflammatory activity of Ju-Zhi-Jiang-Tang, an ancient traditional Chinese medicine formula.
Identification of the effective constituents for anti-inflammatory activity of Ju-Zhi-Jiang-Tang, an ancient traditional Chinese medicine formula.
Journal of chromatography. A (2014-05-13)
Shufang Wang, Pinghong Chen, Wei Jiang, Leihong Wu, Lulin Chen, Xiaohui Fan, Yi Wang, Yiyu Cheng
ABSTRACT
The anti-inflammatory constituents of Ju-Zhi-Jiang-Tang (JZJT), a formula used for thousands of years in China, were identified by LC-MS and pharmacological activity evaluation. In this study, the whole extract of formula was separated into multiple components to facilitate the analytical process. To characterize their contributions to pharmacological activity of formula, activity indexes of constituents were proposed and calculated for the first time, which integrated the chemical and pharmacological information of multiple components. Among the 151 constituents detected in JZJT by LC-Q-TOF-MS and LC-IT-MS, a total number of 108 constituents were identified unambiguously or tentatively, including eighteen potential novel compounds. And, the structures of some constituents were confirmed by NMR. According to their activity indexes, polymethoxy flavones were indicated as the major active constituents responsible for the anti-inflammatory activity of JZJT. To verify the feasibility of activity indexes in predicting the active constituents, nine compounds with positive and negative index values were selected to validate their anti-inflammatory activity in vitro. The results showed that two polymethoxy flavones with higher positive index values, i.e., nobiletin and tangeretin can significantly exert anti-inflammatory effects, while other compounds with negative values did not show any activity. In conclusion, our results indicated the proposed approach might be an efficient and rapid way to identify active constituents of TCM formulae.
Product Description
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Neoeriocitrin, ~95% (HPLC)
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Ultrapure Acetonitrile
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis