Merck
  • Nanotentacle-structured magnetic particles for efficient capture of circulating tumor cells.
Nanotentacle-structured magnetic particles for efficient capture of circulating tumor cells.
Small (Weinheim an der Bergstrasse, Germany) (2014-12-17)
Seong-Min Jo, Joong-jae Lee, Woosung Heu, Hak-Sung Kim
ABSTRACT
Circulating tumor cells (CTCs) have attracted considerable attention as promising markers for diagnosing and monitoring the cancer status. Despite many technological advances in isolating CTCs, the capture efficiency and purity still remain challenges that limit clinical practice. Here, the construction of "nanotentacle"-structured magnetic particles using M13-bacteriophage and their application for the efficient capturing of CTCs is demonstrated. The M13-bacteriophage to magnetic particles followed by modification with PEG is conjugated, and further tethered monoclonal antibodies against the epidermal receptor 2 (HER2). The use of nanotentacle-structured magnetic particles results in a high capture purity (>45%) and efficiency (>90%), even for a smaller number of cancer cells (≈25 cells) in whole blood. Furthermore, the cancer cells captured are shown to maintain a viability of greater than 84%. The approach can be effectively used for capturing CTCs with high efficiency and purity for the diagnosis and monitoring of cancer status.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Neodymium, powder, −40 mesh, ≥99% trace rare earth metals basis
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Neodymium, rod, 100mm, diameter 12.5mm, cast, 99%
Neodymium, rod, 50mm, diameter 6.35mm, cast, 99%
Neodymium, rod, 100mm, diameter 6.35mm, cast, 99%
Neodymium, rod, 50mm, diameter 12.5mm, cast, 99%
Sigma-Aldrich
4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt, powder
Sigma-Aldrich
Propidium iodide solution