Merck
  • Flavonoid composition and antioxidant activities of Chinese local pummelo (Citrus grandis Osbeck.) varieties.
Flavonoid composition and antioxidant activities of Chinese local pummelo (Citrus grandis Osbeck.) varieties.
Food chemistry (2014-05-20)
Wanpeng Xi, Bo Fang, Qiyang Zhao, Bining Jiao, Zhiqin Zhou
ABSTRACT
China is one of the most important diversity centres of the genus Citrus L. and is particularly rich in pummelo germplasm. In this study, the flavonoids in the peels and pulps of 28 Chinese local pummelos and four grapefruits were determined by optimised Ultra Performance Liquid Chromatography (UPLC), and their antioxidant capacities were evaluated using 2,2-diphenyl-1-picrylhydrazyl radicals (DPPH), ferric reducing/antioxidant power (FRAP) and superoxide anion methods. We found that naringin was the predominant flavonoid in pummelo, while naringin and neohesperidin were the predominant flavonoids in grapefruit. The fruit peels of Citrus paradisi cvs. Rio Red and Cocktail had the highest contents of naringin (9871.69mg/kg FW) and neohesperidin (7011.15mg/kg FW), respectively. Overall, C. paradisi cvs. Cocktail, Rio Red and Changshanhuyou, Citrus grandis cvs. 28-19, Chandler, and Hongxinyou contained more flavonoids and exhibited higher antioxidant capacities and are potentially good sources of phytochemicals and natural antioxidants.
Product Description
Supelco
Acetic acid, analytical standard
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. (of Fe), ≥98%
Sigma-Aldrich
(±)-Naringenin, ≥95%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Naringenin, natural (US), 98%
Eriocitrin, primary reference standard
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Nobiletin, ≥97%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Supelco
(±)-Naringenin, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Naringin, ≥95% (HPLC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Hesperidin, ≥80% (HPLC)
Supelco
Hesperetin, analytical standard