Merck
  • Hyaluronic acid-tumor necrosis factor-related apoptosis-inducing ligand conjugate for targeted treatment of liver fibrosis.
Hyaluronic acid-tumor necrosis factor-related apoptosis-inducing ligand conjugate for targeted treatment of liver fibrosis.
Acta biomaterialia (2014-10-12)
Jeong-A Yang, Won Ho Kong, Dong Kyung Sung, Hyemin Kim, Tae Hyung Kim, Kang Choon Lee, Sei Kwang Hahn
ABSTRACT
Liver fibrosis is a chronic liver disease caused by viral infection and/or metabolic, genetic and cholestatic disorders. The inhibition of hepatic stellate cell (HSC) activation and the selective apoptosis of activated HSCs can be a good strategy to treat liver fibrosis. The activated HSCs are known to be more susceptible to tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) induced apoptosis than normal HSCs because death receptor 5 is overexpressed on the cell surface. In this work, a target-specific and long-acting hyaluronic acid (HA)-TRAIL conjugate was successfully developed for the treatment of liver fibrosis. The HA-TRAIL conjugate was synthesized by a coupling reaction between aldehyde-modified HA and the N-terminal amine group of TRAIL. The biological activity of the HA-TRAIL conjugate was confirmed by an in vitro anti-proliferation assay and caspase-3 expression in human colon cancer HCT116 cells. In vivo real-time bioimaging exhibited the target-specific delivery of near-infrared fluorescence dye-labeled HA-TRAIL conjugate to the liver in mice. According to pharmacokinetic analysis, the HA-TRAIL conjugate was detected for more than 4days after single intravenous injection into Sprague-Dawley (SD) rats. Finally, we could confirm the antifibrotic effect of HA-TRAIL conjugate in an N-nitrosodimethylamine-induced liver fibrosis model SD rats.
Sigma-Aldrich
tert-Butyl carbazate, 98%
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium (meta)periodate, ≥99.0%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Sodium periodate, ACS reagent, ≥99.8%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%