Merck
  • The effect of salsalate therapy on endothelial function in a broad range of subjects.
The effect of salsalate therapy on endothelial function in a broad range of subjects.
Journal of the American Heart Association (2014-01-07)
Anju Nohria, Scott Kinlay, J Stewart Buck, Whitney Redline, Robert Copeland-Halperin, Sora Kim, Joshua A Beckman
ABSTRACT
Inflammation is fundamental to the development of atherosclerosis. We examined the effect of anti-inflammatory doses of salicylate on endothelium-dependent vasodilation, a biomarker of cardiovascular risk, in a broad range of subjects. We performed a randomized, double-blind, placebo-controlled crossover trial evaluating the effects of 4 weeks of high-dose salsalate (disalicylate) therapy on endothelium-dependent flow-mediated and endothelium-independent vasodilation. Fifty-eight subjects, including 17 with metabolic syndrome, 13 with atherosclerosis, and 28 healthy controls, were studied. Among all subjects, endothelium-dependent flow-mediated vasodilation decreased after salsalate compared with placebo therapy (P=0.01), whereas nitroglycerin-mediated, endothelium-independent vasodilation was unchanged (P=0.97). Endothelium-dependent flow-mediated vasodilation after salsalate therapy was impaired compared with placebo therapy in subjects with therapeutic salicylate levels (n=31, P<0.02) but not in subjects with subtherapeutic levels (P>0.2). Salsalate therapy, particularly when therapeutic salicylate levels are achieved, impairs endothelium-dependent vasodilation in a broad range of subjects. These data raise concern about the possible deleterious effects of anti-inflammatory doses of salsalate on cardiovascular risk. www.clinicaltrials.gov. Unique Identifiers: NCT00760019 and NCT00762827.
Product Description
Supelco
Omeprazole, analytical standard
Omeprazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Omeprazole, solid
Supelco
Omeprazole, Pharmaceutical Secondary Standard; Certified Reference Material
Omeprazole, European Pharmacopoeia (EP) Reference Standard
USP
Omeprazole, United States Pharmacopeia (USP) Reference Standard