Merck
The crystal structure of arginyl-tRNA synthetase from Homo sapiens.
FEBS letters (2014-05-27)
Hyun Sook Kim, So Young Cha, Chang Hwa Jo, Ahreum Han, Kwang Yeon Hwang
ABSTRACT
Arginyl-tRNA synthetase (ArgRS) is a tRNA-binding protein that catalyzes the esterification of L-arginine to its cognate tRNA. L-Canavanine, a structural analog of L-arginine, has recently been studied as an anticancer agent. Here, we determined the crystal structures of the apo, L-arginine-complexed, and L-canavanine-complexed forms of the cytoplasmic free isoform of human ArgRS (hArgRS). Similar interactions were formed upon binding to L-canavanine or L-arginine, but the interaction between Tyr312 and the oxygen of the oxyguanidino group was a little bit different. Detailed conformational changes that occur upon substrate binding were explained. The hArgRS structure was also compared with previously reported homologue structures. The results presented here may provide a basis for the design of new anticancer drugs, such as L-canavanine analogs.
Sigma-Aldrich
L-Canavanine, ≥98% (TLC), powder, from Canavalia ensiformis
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
SAFC
L-Arginine