Merck
  • Severe infantile encephalomyopathy caused by a mutation in COX6B1, a nucleus-encoded subunit of cytochrome c oxidase.
Severe infantile encephalomyopathy caused by a mutation in COX6B1, a nucleus-encoded subunit of cytochrome c oxidase.
American journal of human genetics (2008-05-24)
Valeria Massa, Erika Fernandez-Vizarra, Saad Alshahwan, Eman Bakhsh, Paola Goffrini, Ileana Ferrero, Paolo Mereghetti, Pio D'Adamo, Paolo Gasparini, Massimo Zeviani
ABSTRACT
Cytochrome c oxidase (COX) deficiency, one of the most common respiratory-chain defects in humans, has been associated with mutations in either mitochondrial DNA genes or nucleus-encoded proteins that are not part in but promote the biogenesis of COX. Mutations of nucleus-encoded structural subunits were sought for but never found in COX-defective patients, leading to the conjecture that they may be incompatible with extra-uterine survival. We report a disease-associated mutation in one such subunit, COX6B1. Nuclear-encoded COX genes should be reconsidered and included in the diagnostic mutational screening of human disorders related to COX deficiency.