Merck
  • An LC-MS/MS method for steroid profiling during adrenal venous sampling for investigation of primary aldosteronism.
An LC-MS/MS method for steroid profiling during adrenal venous sampling for investigation of primary aldosteronism.
The Journal of steroid biochemistry and molecular biology (2014-10-15)
Mirko Peitzsch, Tanja Dekkers, Matthias Haase, Fred C G J Sweep, Ivo Quack, Gerald Antoch, Gabriele Siegert, Jacques W M Lenders, Jaap Deinum, Holger S Willenberg, Graeme Eisenhofer
ABSTRACT
Steroid profiling for diagnosis of endocrine disorders featuring disordered production of steroid hormones is now possible from advances in liquid chromatography with tandem mass spectrometry (LC-MS/MS). Adrenal venous (AV) measurements of aldosterone and cortisol are a standard practice in the clinical work-up of primary aldosteronism, but do not yet take advantage of steroid profiling. A novel LC-MS/MS based method was developed for simultaneous measurement of 15 adrenal steroids: aldosterone, corticosterone, 11-deoxycorticosterone, progesterone, pregnenolone, cortisone, cortisol, 11-deoxycortisol, 17-hydroxyprogesterone, androstenedione, dehydroepiandrosterone, dehydroepiandrosterone-sulfate, 21-deoxycortisol, 18-oxocortisol and 18-hydroxycortisol. These were compared in peripheral venous (pV) and AV plasma from 70 patients undergoing AV sampling with and without cosyntropin stimulation. Aldosterone and cortisol levels measured by LC-MS/MS were compared with those measured by immunoassay. Reproducibility of measurements with coefficients of variation ≤10% as well as analytical sensitivity sufficient to measure low pV levels particularly of aldosterone demonstrate the utility of the assay for profiling adrenal steroids in primary aldosteronism. Method comparisons indicated assay and concentration dependent differences of cortisol and aldosterone concentrations measured by immunoassay and LC-MS/MS. Median AV/pV ratios of 11-deoxycortisol (53.0), 17-hydroxyprogesterone (33.4), pregnenolone (62.4), androstenedione (40.6) and dehydroepiandrosterone (33.3) were 2.9- to, 5.4-fold larger than those for cortisol (11.6), with additionally generally larger increases than for cortisol with than without cosyntropin stimulation. Our LC-MS/MS assay, in addition to improvements over existing immunoassay measurements of aldosterone and cortisol, offers profiling of 13 other adrenal steroids, providing a potentially useful method for the clinical work-up of patients with primary aldosteronism. In particular, the larger AV/pV ratios of several steroids compared to cortisol suggest more sensitive alternatives to the latter for assessing positioning of AV sampling catheters.
Product Description
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
11-Deoxycortisol-2,2,4,6,6-d5, 98 atom % D, 98% (CP)
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard