Merck
  • T-cell variant of classical Hodgkin's lymphoma with nodal and cutaneous manifestations demonstrated by single-cell polymerase chain reaction.
T-cell variant of classical Hodgkin's lymphoma with nodal and cutaneous manifestations demonstrated by single-cell polymerase chain reaction.
Laboratory investigation; a journal of technical methods and pathology (2002-09-10)
Klaus Willenbrock, Ryo Ichinohasama, Marshall E Kadin, Ikuo Miura, Tadashi Terui, Kuniaki Meguro, Osamu Fukuhara, John F DeCoteau, Martin-Leo Hansmann
ABSTRACT
The atypical cells of CD30(+) cutaneous lymphoproliferative disorders (CD30CLD) are commonly of T-cell origin and frequently have a similar morphology as Hodgkin or Reed-Sternberg cells of Hodgkin's lymphoma (HL). HL is one of the tumors associated with CD30CLD. Although most studies support a B-cell derivation of the tumor cells in HL, recently a few cases of classical HL with T-cell genotype have been reported. We report a patient who presented with CD30CLD whose lymph nodes showed classical HL of mixed cellularity subtype at presentation. By single-cell PCR, the same clonal gene rearrangements of the T cell receptor-beta gene locus could be assigned to the CD30(+) and CD15(+) cells of both skin and lymph node. In a lymph node biopsy specimen taken in relapse after several courses of chemotherapy, the CD30(+) tumor cells were abundant. The T cell-derived tumor cells displayed aberrant expression of the Pax-5 gene in all specimens. A common clonal origin of both CD30CLD and HL of the lymph node in the patient presented here suggests that HL with T-cell genotype exists in association with CD30CLD as well as in sporadic cases and may share clonal origin with the skin tumor.
Sigma-Aldrich
PAX-5 (SP34) Rabbit Monoclonal Antibody
Sigma-Aldrich
PAX-5 (24) Mouse Monoclonal Antibody